Tisk…

Benzyl 2-acetamido-2-deoxy-α-D-glucopyranoside

Dodavatel: Apollo Scientific
Varování

Synonyms: Benzyl 2-(acetylamino)-2-deoxy-α-D-glucopyranoside, N-((2S,3R,4R,5S,6R)-2-(Benzyloxy)-4,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)acetamide

OR1254T-1G OR1254T-250MG OR1254T-5G
APOSOR1254T-1GEA 0 CZK
APOSOR1254T-1G APOSOR1254T-250MG APOSOR1254T-5G
Benzyl 2-acetamido-2-deoxy-α-D-glucopyranoside
Benzyl 2-acetamido-2-deoxy-α-D-glucopyranoside
Vzorec: C₁₅H₂₁NO₆
MW: 311,33 g/mol
Bod tání: 179…182 °C
Storage Temperature: Freezer
MDL Number: MFCD00070371
CAS číslo: 13343-62-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR