Tisk…

2-Isobutylpyrrolidine hydrochloride 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Isobutyltetrahydropyrrole hydrochloride, 2-(2-Methylprop-1-yl)pyrrolidine hydrochloride

OR12520-1G OR12520-5G
APOSOR12520-1GEA 4570 CZK
APOSOR12520-1G APOSOR12520-5G
2-Isobutylpyrrolidine hydrochloride 95%
2-Isobutylpyrrolidine hydrochloride
Vzorec: C₈H₁₇N·ClH
MW: 163.688 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03840008
CAS číslo: 1184994-37-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR