3-Bromo-4-(hydroxymethyl)thiophene 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(4-Bromothien-3-yl)methanol, (4-Bromothiophen-3-yl)methanol
OR12445-1G
OR12445-250MG
APOSOR12445-1GEA
7050
CZK
APOSOR12445-1G
APOSOR12445-250MG
3-Bromo-4-(hydroxymethyl)thiophene 97%
3-Bromo-4-(hydroxymethyl)thiophene
Vzorec:
C₅H₅BrOS MW: 193,062 g/mol Bod tání: 53…53,5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01859831 CAS číslo: 70260-05-8 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...