Methyl 4-hydroxyquinazoline-7-carboxylate 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Hydroxy-7-(methoxycarbonyl)quinazoline, 4-Hydroxy-7-(methoxycarbonyl)-1,3-benzodiazine
OR12426-1G
APOSOR12426-1GEA
2070
CZK
APOSOR12426-1G
Methyl 4-hydroxyquinazoline-7-carboxylate 97%
Methyl 4-hydroxyquinazoline-7-carboxylate
Vzorec:
C₁₀H₈N₂O₃ MW: 204,182 g/mol Bod tání: 252…254 °C Storage Temperature: Ambient |
MDL Number:
MFCD00681283 CAS číslo: 313535-84-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...