Tisk…

5-(Aminomethyl)benzo[b]furan 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 5-(Aminomethyl)-1-benzofuran, (Benzo[b]furan-5-yl)methylamine

OR12314-1G OR12314-250MG
APOSOR12314-1GEA 9860 CZK
APOSOR12314-1G APOSOR12314-250MG
5-(Aminomethyl)benzo[b]furan 97%
5-(Aminomethyl)benzo[b]furan
Vzorec: C₉H₉NO
MW: 147,174 g/mol
Bod tání: 32…34 °C
Storage Temperature: Ambient
MDL Number: MFCD04974039
CAS číslo: 37798-08-6
UN: 3259
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR