Tisk…

2-(4-Isocyanatophenyl)pyrimidine

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4-(Pyrimidin-2-yl)phenyl isocyanate, 2-(4-Isocyanatophenyl)-1,3-diazine

OR12304-1G OR12304-250MG
APOSOR12304-1GEA 8100 CZK
APOSOR12304-1G APOSOR12304-250MG
2-(4-Isocyanatophenyl)pyrimidine
2-(4-Isocyanatophenyl)pyrimidine

When melted, may contain small amount of sediment

Vzorec: C₁₁H₇N₃O
MW: 197,193 g/mol
Bod tání: 85…88 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09702380
CAS číslo: 216059-84-6
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR