5-Bromo-1,2-dimethyl-1H-imidazole 96%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR1220-5G
APOSOR1220-5GEA
3380
CZK
APOSOR1220-5G
5-Bromo-1,2-dimethyl-1H-imidazole 96%
5-Bromo-1,2-dimethyl-1H-imidazole
Vzorec:
C₅H₇BrN₂ MW: 175,026 g/mol Bod tání: 91…94 °C Storage Temperature: Ambient |
MDL Number:
MFCD06659905 CAS číslo: 24134-09-6 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...