Tisk…
4-(3-Chloroprop-1-yl)morpholine hydrochloride solution 65% in toluene
Dodavatel: Apollo Scientific
Nebezpečí
Synonyms: 4-(3-Chloropropyl)morpholine hydrochloride, 1-Chloro-3-(morpholin-4-yl)propane hydrochloride
OR11729-1G OR11729-5G
APOSOR11729-1GEA 3310 CZK
APOSOR11729-1G APOSOR11729-5G
4-(3-Chloroprop-1-yl)morpholine hydrochloride solution 65% in toluene
4-(3-Chloroprop-1-yl)morpholine hydrochloride
Vzorec: C₇H₁₄ClNO·ClH
MW: 200.106 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00035333
CAS číslo: 57616-74-7
EINECS: 260-851-0
UN: 3286
ADR: 3+6.1+8 II