Methyl 5-bromo-4-methylthiophene-2-carboxylate 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Methyl 5-bromo-4-methyl-2-thenoate, 5-Bromo-2-(methoxycarbonyl)-4-methylthiophene
OR11283-1G
OR11283-250MG
OR11283-5G
APOSOR11283-1GEA
2570
CZK
APOSOR11283-1G
APOSOR11283-250MG
APOSOR11283-5G
Methyl 5-bromo-4-methylthiophene-2-carboxylate 98%
Methyl 5-bromo-4-methylthiophene-2-carboxylate
Vzorec:
C₇H₇BrO₂S Bod tání: 39…40 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD04038959 CAS číslo: 54796-47-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...