Tisk…

5-Bromo-2,4-dichlorobenzyl alcohol

Dodavatel: Apollo Scientific

Synonyms: (5-Bromo-2,4-dichlorophenyl)methanol

OR111619-1G OR111619-250MG
APOSOR111619-1GEA 10350 CZK
APOSOR111619-1G APOSOR111619-250MG
5-Bromo-2,4-dichlorobenzyl alcohol
5-Bromo-2,4-dichlorobenzyl alcohol
Vzorec: C₇H₅BrCl₂O
MW: 255,924 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28737839
CAS číslo: 1805023-63-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR