Tisk…

Ethyl 3-amino-7-bromo-5-chloro-1-benzofuran-2-carboxylate

Dodavatel: Apollo Scientific

OR111613-1G OR111613-250MG
APOSOR111613-1GEA 8930 CZK
APOSOR111613-1G APOSOR111613-250MG
Ethyl 3-amino-7-bromo-5-chloro-1-benzofuran-2-carboxylate
Ethyl 3-amino-7-bromo-5-chloro-1-benzofuran-2-carboxylate
Vzorec: C₁₁H₉BrClNO₃
Storage Temperature: Ambient
MDL Number: MFCD21609336
CAS číslo: 2183997-56-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR