Tisk…

2-Chloro-3,4-dimethoxy-benzenemethanamine

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: (2-Chloro-3,4-dimethoxyphenyl)methanamine

OR111609-1G OR111609-250MG
APOSOR111609-1GEA 9490 CZK
APOSOR111609-1G APOSOR111609-250MG
2-Chloro-3,4-dimethoxy-benzenemethanamine
2-Chloro-3,4-dimethoxy-benzenemethanamine
Vzorec: C₉H₁₂ClNO₂
MW: 201.65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03410970
CAS číslo: 771580-80-4
UN: 3259
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR