Ethyl (4-bromo-2-cyanophenoxy)acetate
Dodavatel: Apollo Scientific
OR111594-1G
OR111594-5G
APOSOR111594-1GEA
3540
CZK
APOSOR111594-1G
APOSOR111594-5G
Ethyl (4-bromo-2-cyanophenoxy)acetate
Ethyl (4-bromo-2-cyanophenoxy)acetate
Vzorec:
C₁₁H₁₀BrNO₃ Storage Temperature: Ambient |
MDL Number:
MFCD00454355 CAS číslo: 328009-03-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...