Tisk…

Ethyl (2-acetyl-4-bromophenoxy)acetate

Dodavatel: Apollo Scientific

OR111593-1G OR111593-5G
APOSOR111593-1GEA 4240 CZK
APOSOR111593-1G APOSOR111593-5G
Ethyl (2-acetyl-4-bromophenoxy)acetate
Ethyl (2-acetyl-4-bromophenoxy)acetate
Vzorec: C₁₂H₁₃BrO₄
MW: 301,133 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17964567
CAS číslo: 34849-50-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR