Tisk…

7-Iodo-1-benzofuran

Dodavatel: Apollo Scientific
Varování

OR111589-1G OR111589-250MG
APOSOR111589-1GEA 11180 CZK
APOSOR111589-1G APOSOR111589-250MG
7-Iodo-1-benzofuran
7-Iodo-1-benzofuran
Vzorec: C₈H₅IO
MW: 244,029 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20259048
CAS číslo: 1388049-96-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR