Tisk…

(3-Bromo-4-hydroxy-5-nitrophenyl)acetic acid

Dodavatel: Apollo Scientific
Varování

OR111578-1G OR111578-250MG OR111578-5G
APOSOR111578-1GEA 14880 CZK
APOSOR111578-1G APOSOR111578-250MG APOSOR111578-5G
(3-Bromo-4-hydroxy-5-nitrophenyl)acetic acid
(3-Bromo-4-hydroxy-5-nitrophenyl)acetic acid
Vzorec: C₈H₆BrNO₅
MW: 276.041 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32066834
CAS číslo: 38694-44-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR