Tisk…

tert-Butyl 2-cyclobutylhydrazinecarboxylate

Dodavatel: Apollo Scientific
Varování

OR111575-1G OR111575-250MG
APOSOR111575-1GEA 8070 CZK
APOSOR111575-1G APOSOR111575-250MG
tert-Butyl 2-cyclobutylhydrazinecarboxylate
tert-Butyl 2-cyclobutylhydrazinecarboxylate
Vzorec: C₉H₁₈N₂O₂
MW: 186,251 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09991871
CAS číslo: 959137-72-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR