Benzofuran-5-carbonitrile
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR111564-1G
OR111564-5G
APOSOR111564-1GEA
2690
CZK
APOSOR111564-1G
APOSOR111564-5G
Benzofuran-5-carbonitrile
Benzofuran-5-carbonitrile
Vzorec:
C₉H₅NO MW: 143,142 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05664402 CAS číslo: 79002-39-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...