Imidazo[1,2-a]pyridine-6-carbonitrile hydrochloride
Dodavatel: Apollo Scientific
|
Varování
|
OR111553-1G
OR111553-250MG
APOSOR111553-1GEA
705
CZK
APOSOR111553-1G
APOSOR111553-250MG
Imidazo[1,2-a]pyridine-6-carbonitrile hydrochloride
Imidazo[1,2-a]pyridine-6-carbonitrile hydrochloride
Vzorec:
C₈H₅N₃·ClH Storage Temperature: Ambient |
MDL Number:
MFCD31629942 CAS číslo: 1934299-50-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...