Tisk…

6-Methoxy-1H-indazol-3-amine

Dodavatel: Apollo Scientific
Varování

OR111516-1G OR111516-250MG
APOSOR111516-1GEA 8040 CZK
APOSOR111516-1G APOSOR111516-250MG
6-Methoxy-1H-indazol-3-amine
6-Methoxy-1H-indazol-3-amine
Vzorec: C₈H₉N₃O
MW: 163,177 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07385621
CAS číslo: 511225-17-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR