Tisk…

(2-Chloro-3,4-dimethoxyphenyl)acetonitrile

Dodavatel: Apollo Scientific

Synonyms: 2-(2-Chloro-3,4-dimethoxyphenyl)acetonitrile

OR111436-10G OR111436-1G OR111436-5G
APOSOR111436-10GEA 0 CZK
APOSOR111436-10G APOSOR111436-1G APOSOR111436-5G
(2-Chloro-3,4-dimethoxyphenyl)acetonitrile
(2-Chloro-3,4-dimethoxyphenyl)acetonitrile
Vzorec: C₁₀H₁₀ClNO₂
MW: 211,645 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13516980
CAS číslo: 7537-07-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR