Tisk…

4-Chloro-6-(2-ethylpiperidin-1-yl)pyrimidin-2-amine

Dodavatel: Apollo Scientific

OR111392-1G OR111392-5G
APOSOR111392-1GEA 0 CZK
APOSOR111392-1G APOSOR111392-5G
4-Chloro-6-(2-ethylpiperidin-1-yl)pyrimidin-2-amine
4-Chloro-6-(2-ethylpiperidin-1-yl)pyrimidin-2-amine
Vzorec: C₁₁H₁₇ClN₄
MW: 240,732 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23715688
CAS číslo: 1252555-02-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR