Tisk…

Ethyl 3-amino-7-methoxy-1-benzofuran-2-carboxylate

Dodavatel: Apollo Scientific

OR111382-1G OR111382-5G
APOSOR111382-1GEA 6420 CZK
APOSOR111382-1G APOSOR111382-5G
Ethyl 3-amino-7-methoxy-1-benzofuran-2-carboxylate
Ethyl 3-amino-7-methoxy-1-benzofuran-2-carboxylate
Vzorec: C₁₂H₁₃NO₄
MW: 235,236 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20526241
CAS číslo: 1351479-14-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR