4-Methoxy-2,3,6-trimethylbenzenesulphonamide
Dodavatel: Apollo Scientific
OR111377-1G
OR111377-5G
APOSOR111377-1GEA
0
CZK
APOSOR111377-1G
APOSOR111377-5G
4-Methoxy-2,3,6-trimethylbenzenesulphonamide
4-Methoxy-2,3,6-trimethylbenzenesulphonamide
Vzorec:
C₁₀H₁₅NO₃S Storage Temperature: Ambient |
MDL Number:
MFCD30481151 CAS číslo: 316831-31-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...