2-(4-(Chlorosulphonyl)phenoxy)acetic acid
Dodavatel: Apollo Scientific
OR111369-10G
OR111369-1G
OR111369-5G
APOSOR111369-10GEA
0
CZK
APOSOR111369-10G
APOSOR111369-1G
APOSOR111369-5G
2-(4-(Chlorosulphonyl)phenoxy)acetic acid
2-(4-(Chlorosulphonyl)phenoxy)acetic acid
Vzorec:
C₈H₇ClO₅S MW: 250,656 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09947771 CAS číslo: 17641-39-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...