Tisk…

3-Chloro-1-benzothiophene-2-carbonitrile

Dodavatel: Apollo Scientific
Varování

OR111356-1G OR111356-5G OR111356-250MG
APOSOR111356-1GEA 5960 CZK
APOSOR111356-1G APOSOR111356-5G APOSOR111356-250MG
3-Chloro-1-benzothiophene-2-carbonitrile
3-Chloro-1-benzothiophene-2-carbonitrile
Vzorec: C₉H₄ClNS
MW: 193,653 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00631175
CAS číslo: 28989-29-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR