(2-Chloroethyl)cyclohexane
Dodavatel: Apollo Scientific
OR111352-10G
OR111352-1G
OR111352-5G
APOSOR111352-10GEA
0
CZK
APOSOR111352-10G
APOSOR111352-1G
APOSOR111352-5G
(2-Chloroethyl)cyclohexane
(2-Chloroethyl)cyclohexane
Vzorec:
C₈H₁₅Cl Storage Temperature: Ambient |
MDL Number:
MFCD11646261 CAS číslo: 1073-61-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...