Tisk…

3,4-Dichlorobenzyl iodide

Dodavatel: Apollo Scientific
Nebezpečí

OR111348-1G OR111348-5G
APOSOR111348-1GEA 3480 CZK
APOSOR111348-1G APOSOR111348-5G
3,4-Dichlorobenzyl iodide
3,4-Dichlorobenzyl iodide
Vzorec: C₇H₅Cl₂I
Storage Temperature: Ambient
MDL Number: MFCD06410705
CAS číslo: 142523-67-9
UN: 2811
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR