Dimethyl thiophene-2,5-dicarboxylate
Dodavatel: Apollo Scientific
OR111331-1G
OR111331-5G
APOSOR111331-1GEA
0
CZK
APOSOR111331-1G
APOSOR111331-5G
Dimethyl thiophene-2,5-dicarboxylate
Dimethyl thiophene-2,5-dicarboxylate
Vzorec:
C₈H₈O₄S MW: 200,212 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00142121 CAS číslo: 4282-34-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...