Tisk…

Ethyl {4-[(chloroacetyl)amino]phenyl}acetate

Dodavatel: Apollo Scientific

OR111282-1G OR111282-50MG OR111282-250MG OR111282-500MG
APOSOR111282-1GEA 14490 CZK
APOSOR111282-1G APOSOR111282-50MG APOSOR111282-250MG APOSOR111282-500MG
Ethyl {4-[(chloroacetyl)amino]phenyl}acetate
Ethyl {4-[(chloroacetyl)amino]phenyl}acetate
Vzorec: C₁₂H₁₄ClNO₃
MW: 255.697 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08753653
CAS číslo: 928708-29-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR