Tisk…

Ethyl 2,3-dihydro-1-benzofuran-5-ylacetate

Dodavatel: Apollo Scientific

OR111225-1G OR111225-5G
APOSOR111225-1GEA 0 CZK
APOSOR111225-1G APOSOR111225-5G
Ethyl 2,3-dihydro-1-benzofuran-5-ylacetate
Ethyl 2,3-dihydro-1-benzofuran-5-ylacetate
Vzorec: C₁₂H₁₄O₃
MW: 206,238 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11976475
CAS číslo: 69999-18-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR