N-(4-Bromo-2,6-dimethylphenyl)acetamide
Dodavatel: Apollo Scientific
OR111214-10G
OR111214-1G
OR111214-5G
APOSOR111214-10GEA
0
CZK
APOSOR111214-10G
APOSOR111214-1G
APOSOR111214-5G
N-(4-Bromo-2,6-dimethylphenyl)acetamide
N-(4-Bromo-2,6-dimethylphenyl)acetamide
Vzorec:
C₁₀H₁₂BrNO MW: 242,112 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02637614 CAS číslo: 119416-26-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...