Ethyl 3-bromo-4-hydroxy-5-nitrobenzoate
Dodavatel: Apollo Scientific
|
Varování
|
OR111201-1G
OR111201-5G
APOSOR111201-1GEA
2980
CZK
APOSOR111201-1G
APOSOR111201-5G
Ethyl 3-bromo-4-hydroxy-5-nitrobenzoate
Ethyl 3-bromo-4-hydroxy-5-nitrobenzoate
Vzorec:
C₉H₈BrNO₅ MW: 290,068 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22383758 CAS číslo: 141778-85-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...