Tisk…

Ethyl 1-(5-amino-1,3,4-thiadiazol-2-yl)piperidine-3-carboxylate

Dodavatel: Apollo Scientific
Varování

OR111196-1G OR111196-5G
APOSOR111196-1GEA 5920 CZK
APOSOR111196-1G APOSOR111196-5G
Ethyl 1-(5-amino-1,3,4-thiadiazol-2-yl)piperidine-3-carboxylate
Ethyl 1-(5-amino-1,3,4-thiadiazol-2-yl)piperidine-3-carboxylate
Vzorec: C₁₀H₁₆N₄O₂S
MW: 256,325 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29060042
CAS číslo: 2109229-22-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR