3-(Dimethylamino)-1-(2-furyl)prop-2-en-1-one
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-(Dimethylamino)-1-(furan-2-yl)prop-2-en-1-one
OR111190-1G
OR111190-5G
APOSOR111190-1GEA
622
CZK
APOSOR111190-1G
APOSOR111190-5G
3-(Dimethylamino)-1-(2-furyl)prop-2-en-1-one
3-(Dimethylamino)-1-(2-furyl)prop-2-en-1-one
Vzorec:
C₉H₁₁NO₂ MW: 165,189 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00121186 CAS číslo: 17168-45-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...