tert-Butyl 4-(3-cyanobenzyl)piperazine-1-carboxylate
Dodavatel: Apollo Scientific
OR111185-10G
OR111185-1G
OR111185-5G
APOSOR111185-10GEA
0
CZK
APOSOR111185-10G
APOSOR111185-1G
APOSOR111185-5G
tert-Butyl 4-(3-cyanobenzyl)piperazine-1-carboxylate
tert-Butyl 4-(3-cyanobenzyl)piperazine-1-carboxylate
Vzorec:
C₁₇H₂₃N₃O₂ MW: 301,383 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16620852 CAS číslo: 203047-35-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...