3-(Allyloxy)benzaldehyde
Dodavatel: Apollo Scientific
|
Varování
|
OR111184-10G
OR111184-5G
OR111184-1G
APOSOR111184-10GEA
5890
CZK
APOSOR111184-10G
APOSOR111184-5G
APOSOR111184-1G
3-(Allyloxy)benzaldehyde
3-(Allyloxy)benzaldehyde
Vzorec:
C₁₀H₁₀O₂ MW: 162,185 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04227161 CAS číslo: 40359-32-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...