4-(6-Chloroimidazo[1,2-a]pyridin-2-yl)benzonitrile
Dodavatel: Apollo Scientific
OR111168-10G
OR111168-1G
OR111168-5G
APOSOR111168-10GEA
0
CZK
APOSOR111168-10G
APOSOR111168-1G
APOSOR111168-5G
4-(6-Chloroimidazo[1,2-a]pyridin-2-yl)benzonitrile
4-(6-Chloroimidazo[1,2-a]pyridin-2-yl)benzonitrile
Vzorec:
C₁₄H₈ClN₃ MW: 253,686 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22043209 CAS číslo: 118000-64-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...