N'-(2-Bromo-4-isopropylphenyl)-N,N-diisopropylthiourea
Dodavatel: Apollo Scientific
OR111138-1G
OR111138-5G
APOSOR111138-1GEA
0
CZK
APOSOR111138-1G
APOSOR111138-5G
N'-(2-Bromo-4-isopropylphenyl)-N,N-diisopropylthiourea
N'-(2-Bromo-4-isopropylphenyl)-N,N-diisopropylthiourea
Vzorec:
C₁₆H₂₅BrN₂S MW: 357,352 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28987633 CAS číslo: 1924343-26-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...