N,N-Diisopropyl-1H-indole-3-carboxamide
Dodavatel: Apollo Scientific
OR111135-1G
OR111135-5G
APOSOR111135-1GEA
0
CZK
APOSOR111135-1G
APOSOR111135-5G
N,N-Diisopropyl-1H-indole-3-carboxamide
N,N-Diisopropyl-1H-indole-3-carboxamide
Vzorec:
C₁₅H₂₀N₂O MW: 244.332 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD15524847 CAS číslo: 207619-50-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...