Methyl 2-naphthylacetate
Dodavatel: Apollo Scientific
|
Varování
|
OR111126-1G
OR111126-5G
APOSOR111126-1GEA
5790
CZK
APOSOR111126-1G
APOSOR111126-5G
Methyl 2-naphthylacetate
Methyl 2-naphthylacetate
Vzorec:
C₁₃H₁₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD09263745 CAS číslo: 2876-71-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...