Tisk…

1,3-Dimethyl-1H-pyrazolo[3,4-b]pyridine

Dodavatel: Apollo Scientific

OR111121-1G OR111121-5G
APOSOR111121-1GEA 0 CZK
APOSOR111121-1G APOSOR111121-5G
1,3-Dimethyl-1H-pyrazolo[3,4-b]pyridine
1,3-Dimethyl-1H-pyrazolo[3,4-b]pyridine
Vzorec: C₈H₉N₃
MW: 147,177 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12022285
CAS číslo: 116834-97-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR