N,N-Dimethyl-4-nitrobenzamide
Dodavatel: Apollo Scientific
|
Varování
|
OR111112-1G
OR111112-5G
APOSOR111112-1GEA
1700
CZK
APOSOR111112-1G
APOSOR111112-5G
N,N-Dimethyl-4-nitrobenzamide
N,N-Dimethyl-4-nitrobenzamide
Vzorec:
C₉H₁₀N₂O₃ MW: 194,187 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00229078 CAS číslo: 7291-01-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...