Tisk…

Methyl thieno[3,2-b]pyridine-2-carboxylate

Dodavatel: Apollo Scientific
Varování

OR111087-1G OR111087-100MG OR111087-250MG
APOSOR111087-1GEA 13230 CZK
APOSOR111087-1G APOSOR111087-100MG APOSOR111087-250MG
Methyl thieno[3,2-b]pyridine-2-carboxylate
Methyl thieno[3,2-b]pyridine-2-carboxylate
Vzorec: C₉H₇NO₂S
Storage Temperature: Ambient
MDL Number: MFCD12022282
CAS číslo: 478149-02-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR