Tisk…

2-[(5-Amino-1,3,4-thiadiazol-2-yl)amino]ethanol

Dodavatel: Apollo Scientific
Varování

OR111084-1G OR111084-5G
APOSOR111084-1GEA 1660 CZK
APOSOR111084-1G APOSOR111084-5G
2-[(5-Amino-1,3,4-thiadiazol-2-yl)amino]ethanol
2-[(5-Amino-1,3,4-thiadiazol-2-yl)amino]ethanol
Vzorec: C₄H₈N₄OS
Storage Temperature: Ambient
MDL Number: MFCD28347985
CAS číslo: 1858241-76-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR