4-Iodo-N-methylbenzenesulphonamide
Dodavatel: Apollo Scientific
|
Varování
|
OR111052-1G
OR111052-5G
APOSOR111052-1GEA
3310
CZK
APOSOR111052-1G
APOSOR111052-5G
4-Iodo-N-methylbenzenesulphonamide
4-Iodo-N-methylbenzenesulphonamide
Vzorec:
C₇H₈INO₂S Storage Temperature: Ambient |
MDL Number:
MFCD11647536 CAS číslo: 134283-64-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...