Methyl 2-morpholin-4-ylbenzoate
Dodavatel: Apollo Scientific
OR111046-10G
OR111046-1G
OR111046-5G
APOSOR111046-10GEA
0
CZK
APOSOR111046-10G
APOSOR111046-1G
APOSOR111046-5G
Methyl 2-morpholin-4-ylbenzoate
Methyl 2-morpholin-4-ylbenzoate
Vzorec:
C₁₂H₁₅NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD06203763 CAS číslo: 223560-37-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...