Tisk…

1-Benzyl-4,6-dimethyl-2-oxo-1,2-dihydro-pyridine-3-carbonitrile

Dodavatel: Apollo Scientific

OR111020-1G OR111020-5G
APOSOR111020-1GEA 7050 CZK
APOSOR111020-1G APOSOR111020-5G
1-Benzyl-4,6-dimethyl-2-oxo-1,2-dihydro-pyridine-3-carbonitrile
1-Benzyl-4,6-dimethyl-2-oxo-1,2-dihydro-pyridine-3-carbonitrile
Vzorec: C₁₅H₁₄N₂O
MW: 238,284 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00221300
CAS číslo: 64488-19-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR