Ethyl 4-methylthiophene-2-carboxylate
Dodavatel: Apollo Scientific
OR111015-1G
OR111015-5G
APOSOR111015-1GEA
4240
CZK
APOSOR111015-1G
APOSOR111015-5G
Ethyl 4-methylthiophene-2-carboxylate
Ethyl 4-methylthiophene-2-carboxylate
Vzorec:
C₈H₁₀O₂S MW: 170,229 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12025317 CAS číslo: 14282-79-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...