3-(Dimethylamino)-1-thien-2-ylprop-2-en-1-one
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-(Dimethylamino)-1-(thiophen-2-yl)prop-2-en-1-one
OR110972-1G
OR110972-5G
OR110972-25G
APOSOR110972-1GEA
1210
CZK
APOSOR110972-1G
APOSOR110972-5G
APOSOR110972-25G
3-(Dimethylamino)-1-thien-2-ylprop-2-en-1-one
3-(Dimethylamino)-1-thien-2-ylprop-2-en-1-one
Vzorec:
C₉H₁₁NOS MW: 181,255 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00172112 CAS číslo: 34772-98-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...